Assay Details
| Gene Symbol: BYSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001085599.4 | XP_001085599.2 | 760 | 82 | ||
| GenBank mRNA | JU336130.1 | - | 451 | 82 | ||
| JU476993.1 | - | 579 | 82 | |||
| JV048081.1 | - | 625 | 82 | |||
| JV638753.1 | - | 517 | 82 | |||
More Information
Set Membership: |
|

BYSL
Genomic Map