Assay Details
| Gene Symbol: LOC717142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015116665.2 | XP_014972151.2 | 1375 | 148 | ||
| XM_015116666.2 | XP_014972152.1 | 1231 | 148 | |||
| XM_015116667.2 | XP_014972153.1 | 1421 | 148 | |||
| XM_015116668.2 | XP_014972154.1 | 1714 | 148 | |||
| XM_028842759.1 | XP_028698592.1 | 1679 | 148 | |||
| GenBank mRNA | JU325791.1 | - | 979 | 148 | ||
| JU473411.1 | - | 1088 | 148 | |||
| JV045533.1 | - | 1074 | 148 | |||
| JV453234.1 | - | 1066 | 148 | |||
| JV527769.1 | - | 1108 | 148 | |||
| JV568008.1 | - | 1144 | 148 | |||
| Gene Symbol: KYAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028834738.1 | XP_028690571.1 | 1687 | 148 | ||
| XM_028834739.1 | XP_028690572.1 | 1390 | 148 | |||
| XM_028834740.1 | XP_028690573.1 | 1250 | 148 | |||
| XM_028834741.1 | XP_028690574.1 | 1486 | 148 | |||
| XM_028834742.1 | XP_028690575.1 | 1135 | 148 | |||
| XM_028834743.1 | XP_028690576.1 | 1146 | 148 | |||
| GenBank mRNA | JV601807.1 | - | 1140 | 148 | ||
More Information
Set Membership: |
|

LOC717142
Genomic Map