Assay Details
| Gene Symbol: COCH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260563.1 | NP_001247492.1 | 590 | 97 | ||
| XM_015143480.2 | XP_014998966.1 | 321 | 97 | |||
| GenBank mRNA | JV043954.1 | - | 590 | 97 | ||
| JV635846.1 | - | 609 | 97 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

COCH
Genomic Map