Assay Details
Gene Symbol: LOC719690 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842789.1 | XP_028698622.1 | 99 | 88 | ||
GenBank mRNA | JU334622.1 | - | 2797 | 88 | ||
JV047134.1 | - | 2824 | 88 | |||
JV652224.1 | - | 2844 | 88 |
Gene Symbol: MYO9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028839737.1 | XP_028695570.1 | 2706 | 88 |
