Assay Details
Gene Symbol: LOC719690 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842789.1 | XP_028698622.1 | 1780 | 97 | ||
GenBank mRNA | JU334622.1 | - | 4478 | 97 | ||
JV047134.1 | - | 4505 | 97 | |||
JV652224.1 | - | 4528 | 97 |
Gene Symbol: LOC114674126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028839738.1 | XP_028695571.1 | 120 | 97 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028839738.1 transcripts.
Set Membership: |
![]() ![]() |
