Assay Details
Gene Symbol: LOC719690 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842789.1 | XP_028698622.1 | 3209 | 118 | ||
GenBank mRNA | JU334622.1 | - | 5910 | 118 | ||
JV047134.1 | - | 5934 | 118 | |||
JV652224.1 | - | 5957 | 118 |
Gene Symbol: LOC114674126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028839738.1 | XP_028695571.1 | 1552 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() |
