Assay Details
| Gene Symbol: NINJ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194415.2 | NP_001181344.1 | 124 | 132 | ||
| XM_015117252.2 | XP_014972738.1 | 460 | 132 | |||
| GenBank mRNA | JU475729.1 | - | 156 | 132 | ||
| JV047169.1 | - | 157 | 132 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NINJ1
Genomic Map