Assay Details
| Gene Symbol: STK19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136036.2 | XP_014991522.2 | 76 | |||
| GenBank mRNA | JV726085.1 | - | 205 | 76 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

STK19| Gene Symbol: STK19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136036.2 | XP_014991522.2 | 76 | |||
| GenBank mRNA | JV726085.1 | - | 205 | 76 | ||
Set Membership: |
|