Assay Details
| Gene Symbol: MFHAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001090936.4 | XP_001090936.3 | 3552 | 112 | ||
| GenBank mRNA | JU477842.1 | - | 3045 | 112 | ||
| JV635419.1 | - | 3109 | 112 | |||
More Information
Set Membership: |
|

MFHAS1
Genomic Map