Assay Details
| Gene Symbol: MED21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258120.1 | NP_001245049.1 | 78 | 74 | ||
| GenBank mRNA | JU473473.1 | - | 78 | 74 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MED21| Gene Symbol: MED21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258120.1 | NP_001245049.1 | 78 | 74 | ||
| GenBank mRNA | JU473473.1 | - | 78 | 74 | ||
Set Membership: |
|