Assay Details
| Gene Symbol: MTRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266680.1 | NP_001253609.1 | 1226 | 87 | ||
| XM_015120894.2 | XP_014976380.1 | 1470 | 87 | |||
| XM_028837108.1 | XP_028692941.1 | 1536 | 87 | |||
| GenBank mRNA | JU337240.1 | - | 1236 | 87 | ||
| JU477952.1 | - | 1239 | 87 | |||
| JV043258.1 | - | 1226 | 87 | |||
More Information
Set Membership: |
|

MTRF1
Genomic Map