Assay Details
| Gene Symbol: CAMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033509.2 | NP_001028681.1 | 314 | 133 | ||
| GenBank mRNA | AF181954.1 | - | 317 | 133 | ||
| JU494369.1 | - | 158 | 133 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CAMP
Genomic Map