Assay Details
| Gene Symbol: FHL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265674.1 | NP_001252603.1 | 366 | 57 | ||
| XM_028845301.1 | XP_028701134.1 | 674 | 57 | |||
| XM_028845302.1 | XP_028701135.1 | 342 | 57 | |||
| GenBank mRNA | JV636032.1 | - | 366 | 57 | ||
More Information
Set Membership: |
|

FHL3
Genomic Map