Assay Details
| Gene Symbol: VEGFD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101077.3 | XP_001101077.2 | 1501 | 123 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

VEGFD| Gene Symbol: VEGFD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101077.3 | XP_001101077.2 | 1501 | 123 | ||
Set Membership: |
|