Assay Details
| Gene Symbol: CARD19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260938.1 | NP_001247867.1 | 440 | 70 | ||
| GenBank mRNA | JU321018.1 | - | 507 | 70 | ||
| JU472062.1 | - | 440 | 70 | |||
| JV637639.1 | - | 536 | 70 | |||
More Information
Set Membership: |
|

CARD19
Genomic Map