Assay Details
| Gene Symbol: LOC696031 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001084683.4 | XP_001084683.3 | 448 | 129 | ||
| XM_028851924.1 | XP_028707757.1 | 517 | 129 | |||
| GenBank mRNA | JU325334.1 | - | 405 | 129 | ||
| JV630940.1 | - | 410 | 129 | |||
| JV636410.1 | - | 425 | 129 | |||
More Information
Set Membership: |
|

LOC696031
Genomic Map