Assay Details
| Gene Symbol: PROK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260651.1 | NP_001247580.1 | 254 | 129 | ||
| GenBank mRNA | JV045378.1 | - | 254 | 129 | ||
| JV636411.1 | - | 247 | 129 | |||
More Information
Set Membership: |
|

PROK1
Genomic Map