Assay Details
| Gene Symbol: EFS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001107716.4 | XP_001107716.2 | 1770 | 80 | ||
| XM_015143405.2 | XP_014998891.2 | 2710 | 80 | |||
| XM_015143406.2 | XP_014998892.2 | 1057 | 80 | |||
| XM_028851430.1 | XP_028707263.1 | 1303 | 80 | |||
| GenBank mRNA | JU332470.1 | - | 1766 | 80 | ||
| JV705061.1 | - | 1383 | 80 | |||
More Information
Set Membership: |
|

EFS
Genomic Map