Assay Details
| Gene Symbol: FAXC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266112.1 | NP_001253041.1 | 701 | 66 | ||
| XM_028846620.1 | XP_028702453.1 | 1137 | 66 | |||
| GenBank mRNA | JU336750.1 | - | 701 | 66 | ||
More Information
Set Membership: |
|

FAXC
Genomic Map