Assay Details
| Gene Symbol: FUT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145151.2 | XP_015000637.1 | 1579 | 85 | ||
| XM_015145152.2 | XP_015000638.1 | 1579 | 85 | |||
| XM_028852561.1 | XP_028708394.1 | 1463 | 85 | |||
| XM_028852562.1 | XP_028708395.1 | 1463 | 85 | |||
| GenBank mRNA | JU329165.1 | - | 284 | 85 | ||
More Information
Set Membership: |
|

FUT10
Genomic Map