Assay Details
| Gene Symbol: KLHL22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261588.1 | NP_001248517.1 | 327 | 91 | ||
| XM_028827145.1 | XP_028682978.1 | 341 | 91 | |||
| GenBank mRNA | JU336780.1 | - | 302 | 91 | ||
| JU477543.1 | - | 296 | 91 | |||
| JV048480.1 | - | 327 | 91 | |||
More Information
Set Membership: |
|

KLHL22
Genomic Map