Assay Details
| Gene Symbol: PWWP3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122340.2 | XP_014977826.1 | 1516 | 124 | ||
| XM_015122341.2 | XP_014977827.1 | 162 | 124 | |||
| XM_028840267.1 | XP_028696100.1 | 1564 | 124 | |||
| XM_028840268.1 | XP_028696101.1 | 1556 | 124 | |||
| XM_028840269.1 | XP_028696102.1 | 1533 | 124 | |||
| GenBank mRNA | JU349754.1 | - | 1442 | 124 | ||
| JV484967.1 | - | 1430 | 124 | |||
| JV699251.1 | - | 1451 | 124 | |||
More Information
Set Membership: |
|

PWWP3A
Genomic Map