Assay Details
| Gene Symbol: TBL1Y | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145068.1 | NP_001138540.1 | 1242 | 131 | ||
| XM_015128593.2 | XP_014984079.1 | 1406 | 131 | |||
| XM_015128594.2 | XP_014984080.1 | 1400 | 131 | |||
| XM_028846702.1 | XP_028702535.1 | 1743 | 131 | |||
| XM_028846920.1 | XP_028702753.1 | 1415 | 131 | |||
| XM_028847159.1 | XP_028702992.1 | 1156 | 131 | |||
| XM_028847779.1 | XP_028703612.1 | 1397 | 131 | |||
| XM_028848160.1 | XP_028703993.1 | 1400 | 131 | |||
| XM_028848388.1 | XP_028704221.1 | 1401 | 131 | |||
| XM_028848649.1 | XP_028704482.1 | 1009 | 131 | |||
| XM_028848903.1 | XP_028704736.1 | 1294 | 131 | |||
| XM_028849163.1 | XP_028704996.1 | 999 | 131 | |||
| XM_028849349.1 | XP_028705182.1 | 921 | 131 | |||
| XM_028849586.1 | XP_028705419.1 | 1630 | 131 | |||
| GenBank mRNA | FJ527023.1 | - | 1242 | 131 | ||
| Gene Symbol: TBL1X | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126920.2 | XP_014982406.2 | 1504 | 131 | ||
| XM_028842122.1 | XP_028697955.1 | 1926 | 131 | |||
| XM_028842123.1 | XP_028697956.1 | 1644 | 131 | |||
| XM_028842124.1 | XP_028697957.1 | 1441 | 131 | |||
| XM_028842125.1 | XP_028697958.1 | 1767 | 131 | |||
| XM_028842126.1 | XP_028697959.1 | 1477 | 131 | |||
| XM_028842127.1 | XP_028697960.1 | 1406 | 131 | |||
| XM_028842128.1 | XP_028697961.1 | 1491 | 131 | |||
| XM_028842129.1 | XP_028697962.1 | 1639 | 131 | |||
| XM_028842130.1 | XP_028697963.1 | 1335 | 131 | |||
| GenBank mRNA | JV048069.1 | - | 1753 | 131 | ||
| JV048070.1 | - | 1618 | 131 | |||
| JV659860.1 | - | 1489 | 131 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TBL1Y
Genomic Map