Assay Details
| Gene Symbol: KNSTRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015141933.2 | XP_014997419.2 | 710 | 96 | ||
| XM_028850740.1 | XP_028706573.1 | 712 | 96 | |||
| XM_028850741.1 | XP_028706574.1 | 714 | 96 | |||
| GenBank mRNA | JU495945.1 | - | 440 | 96 | ||
| JV075023.1 | - | 444 | 96 | |||
More Information
Set Membership: |
|

KNSTRN
Genomic Map