Assay Details
| Gene Symbol: RASL10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194639.2 | NP_001181568.1 | 721 | 61 | ||
| XM_028835714.1 | XP_028691547.1 | 3589 | 61 | |||
| GenBank mRNA | JU322297.1 | - | 575 | 61 | ||
| JV566089.1 | - | 714 | 61 | |||
More Information
Set Membership: |
|

RASL10B
Genomic Map