Assay Details
| Gene Symbol: CENPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261531.1 | NP_001248460.1 | 105 | 72 | ||
| GenBank mRNA | JU477793.1 | - | 105 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001261531.1 transcripts.
Set Membership: |
|

CENPL
Genomic Map