Assay Details
| Gene Symbol: SIGLEC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001116366.3 | XP_001116366.2 | 1455 | 85 | ||
| XM_028839116.1 | XP_028694949.1 | 1102 | 85 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SIGLEC12
Genomic Map