Assay Details
| Gene Symbol: CDC14B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001106436.4 | XP_001106436.1 | 1676 | 134 | ||
| XM_015118114.2 | XP_014973600.1 | 1419 | 134 | |||
| XM_015118116.2 | XP_014973602.1 | 1793 | 134 | |||
| XM_028835381.1 | XP_028691214.1 | 1419 | 134 | |||
| XM_028835382.1 | XP_028691215.1 | 1419 | 134 | |||
| XM_028835384.1 | XP_028691217.1 | 1383 | 134 | |||
| GenBank mRNA | JU495382.1 | - | 1253 | 134 | ||
More Information
Set Membership: |
|

CDC14B
Genomic Map