Assay Details
| Gene Symbol: SEPTIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125938.2 | XP_014981424.1 | 1428 | 97 | ||
| GenBank mRNA | JU481673.1 | - | 757 | 97 | ||
| JU481674.1 | - | 732 | 97 | |||
More Information
Set Membership: |
|

SEPTIN1
Genomic Map