Assay Details
| Gene Symbol: SEPTIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125938.2 | XP_014981424.1 | 1341 | 112 | ||
| XM_028840925.1 | XP_028696758.1 | 1332 | 112 | |||
| XM_028840926.1 | XP_028696759.1 | 1232 | 112 | |||
| GenBank mRNA | JU481673.1 | - | 670 | 112 | ||
| JU481674.1 | - | 645 | 112 | |||
More Information
Set Membership: |
|

SEPTIN1
Genomic Map