Assay Details
| Gene Symbol: PANX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150616.2 | XP_015006102.2 | 288 | 79 | ||
| GenBank mRNA | JU330796.1 | - | 224 | 79 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PANX2| Gene Symbol: PANX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150616.2 | XP_015006102.2 | 288 | 79 | ||
| GenBank mRNA | JU330796.1 | - | 224 | 79 | ||
Set Membership: |
|