Assay Details
Gene Symbol: PCMTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260718.1 | NP_001247647.1 | 844 | 141 | ||
XM_015145337.2 | XP_015000823.1 | 865 | 141 | |||
XM_015145338.2 | XP_015000824.1 | 505 | 141 | |||
XM_015145339.2 | XP_015000825.1 | 720 | 141 | |||
XM_015145341.2 | XP_015000827.1 | 370 | 141 | |||
GenBank mRNA | JU322194.1 | - | 829 | 141 | ||
JU471616.1 | - | 802 | 141 | |||
JV044653.1 | - | 844 | 141 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015145338.2, XM_015145339.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
