Assay Details
| Gene Symbol: TSEN15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001109012.4 | XP_001109012.2 | 315 | 158 | ||
| XM_015121996.2 | XP_014977482.1 | 315 | 158 | |||
| XM_015122003.2 | XP_014977489.1 | 315 | 158 | |||
| XM_015122013.2 | XP_014977499.1 | 315 | 158 | |||
| XM_028851357.1 | XP_028707190.1 | 315 | 158 | |||
| GenBank mRNA | JU321424.1 | - | 261 | 158 | ||
| JU474241.1 | - | 251 | 158 | |||
| JV046134.1 | - | 288 | 158 | |||
More Information
Set Membership: |
|

TSEN15
Genomic Map