Assay Details
| Gene Symbol: ZFP91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015114142.2 | XP_014969628.2 | 1603 | 155 | ||
| XM_015114143.2 | XP_014969629.2 | 1278 | 155 | |||
| GenBank mRNA | JU327536.1 | - | 920 | 155 | ||
| JU477333.1 | - | 1233 | 155 | |||
More Information
Set Membership: |
|

ZFP91
Genomic Map