Assay Details
| Gene Symbol: ATP6V1G2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047145.1 | NP_001040610.1 | 184 | 115 | ||
| GenBank mRNA | JU335026.1 | - | 468 | 115 | ||
| JV548098.1 | - | 473 | 115 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ATP6V1G2
Genomic Map