Assay Details
| Gene Symbol: DYNLRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257686.1 | NP_001244615.1 | 189 | 146 | ||
| GenBank mRNA | JU320704.1 | - | 189 | 146 | ||
| JU472814.1 | - | 137 | 146 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001257686.1 transcripts.
Set Membership: |
|

DYNLRB2
Genomic Map