Assay Details
| Gene Symbol: MTSS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028841116.1 | XP_028696949.1 | 1088 | 110 | ||
| XM_028841117.1 | XP_028696950.1 | 888 | 110 | |||
| XM_028841118.1 | XP_028696951.1 | 897 | 110 | |||
| GenBank mRNA | JV633402.1 | - | 904 | 110 | ||
| JV665533.1 | - | 937 | 110 | |||
More Information
Set Membership: |
|

MTSS2
Genomic Map