Assay Details
| Gene Symbol: MTFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015137376.2 | XP_014992862.1 | 4546 | 99 | ||
| XM_015137377.2 | XP_014992863.1 | 4259 | 99 | |||
| XM_015137378.2 | XP_014992864.1 | 914 | 99 | |||
| XM_015137380.2 | XP_014992866.2 | 838 | 99 | |||
| XM_028847148.1 | XP_028702981.1 | 5175 | 99 | |||
| GenBank mRNA | JU474191.1 | - | 1273 | 99 | ||
More Information
Set Membership: |
|

MTFR2
Genomic Map