Assay Details
| Gene Symbol: ACYP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001114543.4 | XP_001114543.1 | 616 | 87 | ||
| XM_028831682.1 | XP_028687515.1 | 358 | 87 | |||
| GenBank mRNA | JV639157.1 | - | 506 | 87 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ACYP2
Genomic Map