Assay Details
| Gene Symbol: C1H1orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015110674.2 | XP_014966160.1 | 889 | 133 | ||
| XM_015110681.2 | XP_014966167.1 | 835 | 133 | |||
| XM_015110691.2 | XP_014966177.1 | 812 | 133 | |||
| XM_028828709.1 | XP_028684542.1 | 520 | 133 | |||
| XM_028828715.1 | XP_028684548.1 | 662 | 133 | |||
| GenBank mRNA | JU319736.1 | - | 775 | 133 | ||
| JU333869.1 | - | 721 | 133 | |||
| JU471218.1 | - | 758 | 133 | |||
| JV046682.1 | - | 709 | 133 | |||
| JV636653.1 | - | 828 | 133 | |||
| JV711518.1 | - | 718 | 133 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

C1H1orf43
Genomic Map