Assay Details
| Gene Symbol: CANT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001109215.4 | XP_001109215.2 | 967 | 86 | ||
| XM_015120488.2 | XP_014975974.1 | 1049 | 86 | |||
| GenBank mRNA | JU474285.1 | - | 850 | 86 | ||
More Information
Set Membership: |
|

CANT1
Genomic Map