Assay Details
Gene Symbol: C3H7orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015133839.2 | XP_014989325.2 | 783 | 125 | ||
XM_028845944.1 | XP_028701777.1 | 783 | 125 | |||
XM_028845945.1 | XP_028701778.1 | 783 | 125 |

More Information
Set Membership: |
![]() ![]() |
