Assay Details
| Gene Symbol: TSPAN32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015113358.2 | XP_014968844.2 | 907 | 170 | ||
| XM_028833053.1 | XP_028688886.1 | 910 | 170 | |||
| XM_028833054.1 | XP_028688887.1 | 826 | 170 | |||
| XM_028833055.1 | XP_028688888.1 | 740 | 170 | |||
| XM_028833056.1 | XP_028688889.1 | 826 | 170 | |||
| XM_028833057.1 | XP_028688890.1 | 740 | 170 | |||
| XM_028833058.1 | XP_028688891.1 | 1645 | 170 | |||
More Information
Set Membership: |
|

TSPAN32
Genomic Map