Assay Details
| Gene Symbol: DENND2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015114907.2 | XP_014970393.1 | 3413 | 122 | ||
| XM_015114908.2 | XP_014970394.1 | 3436 | 122 | |||
| XM_015114910.2 | XP_014970396.1 | 3141 | 122 | |||
| XM_015114911.2 | XP_014970397.1 | 3174 | 122 | |||
| XM_015114913.2 | XP_014970399.1 | 3307 | 122 | |||
| XM_015114914.2 | XP_014970400.1 | 3593 | 122 | |||
| XM_015114916.2 | XP_014970402.1 | 2096 | 122 | |||
| XM_015114917.2 | XP_014970403.1 | 2295 | 122 | |||
| XM_015114918.2 | XP_014970404.1 | 2022 | 122 | |||
| XM_015114919.2 | XP_014970405.1 | 1879 | 122 | |||
| XM_028833712.1 | XP_028689545.1 | 2946 | 122 | |||
| XM_028833713.1 | XP_028689546.1 | 3325 | 122 | |||
| XM_028833714.1 | XP_028689547.1 | 3266 | 122 | |||
| XM_028833715.1 | XP_028689548.1 | 3132 | 122 | |||
| XM_028833717.1 | XP_028689550.1 | 3284 | 122 | |||
| XM_028833718.1 | XP_028689551.1 | 2947 | 122 | |||
| XM_028833719.1 | XP_028689552.1 | 1683 | 122 | |||
| GenBank mRNA | JU336078.1 | - | 3090 | 122 | ||
| JV048041.1 | - | 3059 | 122 | |||
| JV048042.1 | - | 3068 | 122 | |||
| JV636336.1 | - | 1823 | 122 | |||
More Information
Set Membership: |
|

DENND2B
Genomic Map