Assay Details
| Gene Symbol: DENND2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015114907.2 | XP_014970393.1 | 3558 | 111 | ||
| XM_015114908.2 | XP_014970394.1 | 3581 | 111 | |||
| XM_015114910.2 | XP_014970396.1 | 3286 | 111 | |||
| XM_015114911.2 | XP_014970397.1 | 3319 | 111 | |||
| XM_015114913.2 | XP_014970399.1 | 3452 | 111 | |||
| XM_015114914.2 | XP_014970400.1 | 3738 | 111 | |||
| XM_015114916.2 | XP_014970402.1 | 2241 | 111 | |||
| XM_015114917.2 | XP_014970403.1 | 2440 | 111 | |||
| XM_015114918.2 | XP_014970404.1 | 2167 | 111 | |||
| XM_015114919.2 | XP_014970405.1 | 2024 | 111 | |||
| XM_028833712.1 | XP_028689545.1 | 3091 | 111 | |||
| XM_028833713.1 | XP_028689546.1 | 3470 | 111 | |||
| XM_028833714.1 | XP_028689547.1 | 3411 | 111 | |||
| XM_028833715.1 | XP_028689548.1 | 3277 | 111 | |||
| XM_028833717.1 | XP_028689550.1 | 3429 | 111 | |||
| XM_028833719.1 | XP_028689552.1 | 1828 | 111 | |||
| GenBank mRNA | JU336078.1 | - | 3235 | 111 | ||
| JV048041.1 | - | 3204 | 111 | |||
| JV048042.1 | - | 3213 | 111 | |||
| JV636336.1 | - | 1968 | 111 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DENND2B
Genomic Map