Assay Details
| Gene Symbol: DENND2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015114907.2 | XP_014970393.1 | 3892 | 123 | ||
| XM_015114908.2 | XP_014970394.1 | 3915 | 123 | |||
| XM_015114910.2 | XP_014970396.1 | 3620 | 123 | |||
| XM_015114911.2 | XP_014970397.1 | 3653 | 123 | |||
| XM_015114913.2 | XP_014970399.1 | 3786 | 123 | |||
| XM_015114914.2 | XP_014970400.1 | 4072 | 123 | |||
| XM_015114916.2 | XP_014970402.1 | 2575 | 123 | |||
| XM_015114917.2 | XP_014970403.1 | 2774 | 123 | |||
| XM_015114918.2 | XP_014970404.1 | 2501 | 123 | |||
| XM_015114919.2 | XP_014970405.1 | 2358 | 123 | |||
| XM_028833712.1 | XP_028689545.1 | 3425 | 123 | |||
| XM_028833713.1 | XP_028689546.1 | 3804 | 123 | |||
| XM_028833714.1 | XP_028689547.1 | 3745 | 123 | |||
| XM_028833715.1 | XP_028689548.1 | 3611 | 123 | |||
| XM_028833717.1 | XP_028689550.1 | 3763 | 123 | |||
| XM_028833719.1 | XP_028689552.1 | 2162 | 123 | |||
| GenBank mRNA | JU336078.1 | - | 3569 | 123 | ||
| JV636336.1 | - | 2302 | 123 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DENND2B
Genomic Map