Assay Details
| Gene Symbol: DENND2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015114907.2 | XP_014970393.1 | 2867 | 86 | ||
| XM_015114908.2 | XP_014970394.1 | 2890 | 86 | |||
| XM_015114910.2 | XP_014970396.1 | 2595 | 86 | |||
| XM_015114911.2 | XP_014970397.1 | 2628 | 86 | |||
| XM_015114913.2 | XP_014970399.1 | 2761 | 86 | |||
| XM_015114914.2 | XP_014970400.1 | 3047 | 86 | |||
| XM_015114916.2 | XP_014970402.1 | 1550 | 86 | |||
| XM_015114917.2 | XP_014970403.1 | 1749 | 86 | |||
| XM_015114918.2 | XP_014970404.1 | 1476 | 86 | |||
| XM_015114919.2 | XP_014970405.1 | 1333 | 86 | |||
| XM_028833712.1 | XP_028689545.1 | 2400 | 86 | |||
| XM_028833713.1 | XP_028689546.1 | 2779 | 86 | |||
| XM_028833714.1 | XP_028689547.1 | 2720 | 86 | |||
| XM_028833715.1 | XP_028689548.1 | 2586 | 86 | |||
| XM_028833717.1 | XP_028689550.1 | 2738 | 86 | |||
| XM_028833718.1 | XP_028689551.1 | 2401 | 86 | |||
| XM_028833719.1 | XP_028689552.1 | 1137 | 86 | |||
| GenBank mRNA | JU336078.1 | - | 2544 | 86 | ||
| JV048041.1 | - | 2513 | 86 | |||
| JV048042.1 | - | 2522 | 86 | |||
| JV636336.1 | - | 1277 | 86 | |||

DENND2B
Genomic Map