Assay Details
| Gene Symbol: FLACC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001097906.4 | XP_001097906.1 | 1099 | 125 | ||
| XM_015110655.2 | XP_014966141.1 | 759 | 125 | |||
| GenBank mRNA | JV707009.1 | - | 169 | 125 | ||
More Information
Set Membership: |
|

FLACC1
Genomic Map