Assay Details
| Gene Symbol: PPM1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131708.2 | XP_014987194.1 | 1449 | 117 | ||
| XM_028843817.1 | XP_028699650.1 | 607 | 117 | |||
| GenBank mRNA | JU333112.1 | - | 820 | 117 | ||
| JU474444.1 | - | 839 | 117 | |||
| JV572450.1 | - | 839 | 117 | |||
More Information
Set Membership: |
|

PPM1L
Genomic Map