Assay Details
| Gene Symbol: ORMDL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193815.1 | NP_001180744.1 | 319 | 118 | ||
| XM_002800455.2 | XP_002800501.1 | 355 | 118 | |||
| XM_015119519.2 | XP_014975005.1 | 1243 | 118 | |||
| GenBank mRNA | JU337165.1 | - | 193 | 118 | ||
| JV048741.1 | - | 295 | 118 | |||
| JV636998.1 | - | 312 | 118 | |||
More Information
Set Membership: |
|

ORMDL3
Genomic Map